5-Methyl-2-thiophenecarboxylic acid hydrazide

Supplier: Matrix Scientific

002210-1G 002210-5G
101783-468EA 64.62 USD
101783-468 101783-470
5-Methyl-2-thiophenecarboxylic acid hydrazide
5-Methyl-2-thiophenecarboxylic acid hydrazide

Hazard: Irritant

Formula: C₆H₈N₂OS
Melting Pt: 100…102 °C
MDL Number: MFCD01310833
CAS Number: 126768-40-9

Order Now

Specification Test Results

Melting point 100-102°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR