5-Chloro-2-thiophenecarboxylic acid hydrazide ≥98%

Supplier: Matrix Scientific

002209-1G 002209-5G
101783-464EA 41.54 USD
101783-464 101783-466
5-Chloro-2-thiophenecarboxylic acid hydrazide ≥98%
5-Chloro-2-thiophenecarboxylic acid hydrazide

Hazard: Irritant

Formula: C₅H₅ClN₂OS
MW: 176.63 g/mol
Melting Pt: 148…149 °C
MDL Number: MFCD01310805
CAS Number: 351983-31-8

Order Now

Specification Test Results

Melting point 148-149°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR