1-(5-Fluoro-2-methylphenyl)-2-thiourea

Supplier: Matrix Scientific

Synonyms: N-(5-Fluoro-2-methylphenyl)thiourea

002181-1G 002181-5G
101783-408EA 73.85 USD
101783-408 101783-410
1-(5-Fluoro-2-methylphenyl)-2-thiourea
1-(5-Fluoro-2-methylphenyl)-2-thiourea

Hazard: Irritant

Formula: C₈H₉FN₂S
MW: 184.23 g/mol
Melting Pt: 125-127°C
MDL Number: MFCD00052380
CAS Number: 16822-86-9
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 125-127°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR