5-Chloro-6-hydroxynicotinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Chloro-6-hydroxy-3-pyridinecarboxylic Acid

002128-1G 002128-5G 002128-25G
101783-270EA 13.85 USD
101783-270 101783-272 101783-274
5-Chloro-6-hydroxynicotinic acid ≥97%
5-Chloro-6-hydroxynicotinic acid

Hazard: Irritant

Formula: C₆H₄ClNO₃
MW: 173.55 g/mol
Melting Pt: >300 °C
MDL Number: MFCD00052950
CAS Number: 54127-63-8

Order Now

Specification Test Results

Melting point >300°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR