5-Chloro-2-hydroxybenzyl alcohol

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-(hydroxymethyl)phenol

002127-1G 002127-5G
101783-266EA 49.23 USD
101783-266 101783-268
5-Chloro-2-hydroxybenzyl alcohol
5-Chloro-2-hydroxybenzyl alcohol

Hazard: Irritant/Refrigerate

Formula: C₇H₇ClO₂
MW: 158.59 g/mol
Melting Pt: 90…91 °C
MDL Number: MFCD00021967
CAS Number: 5330-38-1

Order Now

Specification Test Results

Melting point 90-91°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR