3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

002121-1G 002121-5G
101783-240EA 16.92 USD
101783-240 101783-242
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine ≥98%
3-Chloro-2-hydroxy-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₆H₃ClF₃NO
Melting Pt: 157…159 °C
MDL Number: MFCD00153095
CAS Number: 76041-71-9

Order Now

Specification Test Results

Melting point 157-159°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR