3-Chloro-2-[4-(ethoxycarbonyl)phenoxy]-5-(trifluoromethyl)pyridine

Supplier: Matrix Scientific

002042-250MG 002042-1G
101783-092EA 161.54 USD
101783-092 101783-094
3-Chloro-2-[4-(ethoxycarbonyl)phenoxy]-5-(trifluoromethyl)pyridine
3-Chloro-2-[4-(ethoxycarbonyl)phenoxy]-5-(trifluoromethyl)pyridine

Hazard: Irritant

Formula: C₁₅H₁₁ClF₃NO₃
Melting Pt: 58…60 °C
MDL Number: MFCD00084952
CAS Number: 105626-86-6

Order Now

Specification Test Results

Melting point 58-60°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR