trans-3-Methoxy-β-nitrostyrene ≥98%
Supplier: Matrix Scientific
001877-5G
001877-25G
101782-788EA
106.15
USD
101782-788
101782-790
trans-3-Methoxy-β-nitrostyrene ≥98%
trans-3-Methoxy-β-nitrostyrene
Hazard: Irritant/Refrigerate
Formula:
C₉H₉NO₃ MW: 179.18 g/mol Melting Pt: 93…95 °C |
MDL Number:
MFCD00024822 CAS Number: 3179-09-7 |
Specification Test Results
Melting point | 93-95°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...