4-Methoxy-β-nitrostyrene ≥98%
Supplier: Matrix Scientific
001874-5G
001874-25G
101782-778EA
146.15
USD
101782-778
101782-780
4-Methoxy-β-nitrostyrene ≥98%
4-Methoxy-β-nitrostyrene
Hazard: Irritant/Refrigerate
Formula:
C₉H₉NO₃ Melting Pt: 86…88 °C |
MDL Number:
MFCD00024826 CAS Number: 3179-10-0 |
Specification Test Results
Melting point | 86-88°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...