2-Amino-N-tert-butylbenzamide

Supplier: Matrix Scientific

001867-1G 001867-5G
101782-766EA 60 USD
101782-766 101782-768
2-Amino-N-tert-butylbenzamide
2-Amino-N-tert-butylbenzamide

Hazard: Irritant

Formula: C₁₁H₁₆N₂O
Melting Pt: 124-125°C
MDL Number: MFCD00060601
CAS Number: 1203-89-0

Order Now

Specification Test Results

Melting point 124-125°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR