3-Amino-4-methylbenzotrifluoride

Supplier: Matrix Scientific

Synonyms: 2-Methyl-5-(trifluoromethyl)aniline

001813-5G 001813-25G
101782-652EA 110.77 USD
101782-652 102663-544
3-Amino-4-methylbenzotrifluoride
3-Amino-4-methylbenzotrifluoride

Hazard: Toxic

Formula: C₈H₈F₃N
MW: 175.16 g/mol
Melting Pt: 34-38°C
MDL Number: MFCD00134580
CAS Number: 25449-96-1
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 34-38°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR