3-Amino-2-methylbenzotrifluoride ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Methyl-3-(trifluoromethyl)aniline

001812-5G 001812-25G
101782-646EA 15.38 USD
101782-646 101782-648
3-Amino-2-methylbenzotrifluoride ≥98%
3-Amino-2-methylbenzotrifluoride

Hazard: Toxic

Formula: C₈H₈F₃N
MW: 175.16 g/mol
Boiling Pt: 62-64°C/4mm
Melting Pt: 38-42°C
MDL Number: MFCD00153216
CAS Number: 54396-44-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 38-42°CC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR