5-Bromo-2-methoxyphenylacetonitrile ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(5-Bromo-2-methoxyphenyl)acetonitrile

001799-1G 001799-5G 001799-25G
101782-614EA 72.31 USD
101782-614 101782-616 101782-618
5-Bromo-2-methoxyphenylacetonitrile ≥98%
5-Bromo-2-methoxyphenylacetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₉H₈BrNO
MW: 226.08 g/mol
Melting Pt: 61…63 °C
MDL Number: MFCD00016392
CAS Number: 7062-40-0

Order Now

Specification Test Results

Melting point 61-63°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR