4,4'-(Hexafluoroisopropylidene)diphenol ≥95%

Supplier: Matrix Scientific

001770-100G 001770-500G
101782-526EA 75.38 USD
101782-526 101782-528
4,4'-(Hexafluoroisopropylidene)diphenol ≥95%
4,4'-(Hexafluoroisopropylidene)diphenol

TCA listed: T
Hazard: Irritant

Formula: C₁₅H₁₀F₆O₂
MW: 336.24 g/mol
Melting Pt: 161…162 °C
MDL Number: MFCD00000439
CAS Number: 1478-61-1

Order Now

Specification Test Results

Melting point 161-162°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR