2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine ≥97%

Supplier: Matrix Scientific

Synonyms: 2-Amino-4-hydroxy-6-trifluoromethylpyrimidine

001765-1G 001765-5G 001765-25G
101782-514EA 30.77 USD
101782-514 101782-516 101782-518
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine ≥97%
2-Amino-4-hydroxy-6-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₅H₄F₃N₃O
MW: 179.1 g/mol
Melting Pt: 288…292 °C
MDL Number: MFCD00068093
CAS Number: 1513-69-5

Order Now

Specification Test Results

Melting point 288-292°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR