3-Bromo-4-hydroxy-5-methoxyphenylacetonitrile

Supplier: Matrix Scientific

001764-1G 001764-5G
101782-510EA 158.46 USD
101782-510 101782-512
3-Bromo-4-hydroxy-5-methoxyphenylacetonitrile
3-Bromo-4-hydroxy-5-methoxyphenylacetonitrile

Hazard: Irritant

Formula: C₉H₈BrNO₂
Melting Pt: 145…147 °C
MDL Number: MFCD00060307
CAS Number: 81038-44-0

Order Now

Specification Test Results

Melting point 145-147°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR