4-Amino-5-fluoroquinazoline ≥98%

Supplier: Matrix Scientific

001752-1G 001752-5G
101782-474EA 392.31 USD
101782-474 101782-476
4-Amino-5-fluoroquinazoline ≥98%
4-Amino-5-fluoroquinazoline

Hazard: Irritant

Formula: C₈H₆FN₃
Melting Pt: 212-214°C
MDL Number: MFCD00153064
CAS Number: 137553-48-1

Order Now

Specification Test Results

Melting point 212-214°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR