Ethyl-1,3-dimethyl-4-(4-fluorophenoxy)-1H-pyrazolo[3,4-b]pyridine-5-carboxylate
Supplier: Matrix Scientific
001670-250MG
001670-1G
101782-230EA
73.85
USD
101782-230
101782-232
Ethyl-1,3-dimethyl-4-(4-fluorophenoxy)-1H-pyrazolo[3,4-b]pyridine-5-carboxylate
Ethyl-1,3-dimethyl-4-(4-fluorophenoxy)-1H-pyrazolo[3,4-b]pyridine-5-carboxylate
Hazard: Irritant
Formula:
C₁₇H₁₆FN₃O₃ Melting Pt: 110…111.5 °C |
MDL Number:
MFCD00067965 CAS Number: 174842-35-4 |
Specification Test Results
Melting point | 110-111.5°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...