Ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate

Supplier: Matrix Scientific

Synonyms: Ethyl 1,4-dihydro-8-fluoro-4-oxoquinoline-3-carboxylate

001669-1G 001669-5G
101782-226EA 38.46 USD
101782-226 101782-228
Ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate
Ethyl 8-fluoro-4-hydroxyquinoline-3-carboxylate

Hazard: Irritant

Formula: C₁₂H₁₀FNO₃
MW: 235.211 g/mol
Melting Pt: 178°C
MDL Number: MFCD00052242
CAS Number: 63010-69-5

Order Now

Specification Test Results

Melting point 217-219°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR