(2-Bromo-5-chlorophenyl)acetic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 2-(2-Bromo-5-chlorophenyl)acetic acid

001647-5G 001647-25G
101782-174EA 50.77 USD
101782-174 101782-176
(2-Bromo-5-chlorophenyl)acetic acid ≥98%
(2-Bromo-5-chlorophenyl)acetic acid

Hazard: Irritant

Formula: C₈H₆BrClO₂
MW: 249.4900 g/mol
Melting Pt: 101-103°C
MDL Number: MFCD00153890
CAS Number: 81682-38-4

Order Now

Specification Test Results

Melting point 101-103°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR