5-Amino-4-bromo-1-methyl-1H-pyrazole ≥96%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1-methyl-1H-pyrazol-5-amine

001584-1G 001584-5G
101782-022EA 72.31 USD
101782-022 101782-024
5-Amino-4-bromo-1-methyl-1H-pyrazole ≥96%
5-Amino-4-bromo-1-methyl-1H-pyrazole

Hazard: Irritant

Formula: C₄H₆BrN₃
MW: 176.02 g/mol
Melting Pt: 103…104 °C
MDL Number: MFCD00068157
CAS Number: 105675-85-2

Order Now

Specification Test Results

Melting point 103-104°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR