5-Cumyl-o-anisidine hydrochloride

Supplier: Matrix Scientific

001582-1G 001582-5G
101782-012EA 40 USD
101782-012 101782-014
5-Cumyl-o-anisidine hydrochloride
5-Cumyl-o-anisidine hydrochloride

Hazard: Irritant

Formula: C₁₆H₂₀ClNO
Melting Pt: 173°C
MDL Number: MFCD00270101
CAS Number: 58999-69-2

Order Now

Specification Test Results

Melting point 180-182°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR