5-Cyano-1-[3-(trifluoromethyl)phenyl]uracil
Supplier: Matrix Scientific
001576-1G
001576-5G
101781-998EA
100
USD
101781-998
101782-000
5-Cyano-1-[3-(trifluoromethyl)phenyl]uracil
5-Cyano-1-[3-(trifluoromethyl)phenyl]uracil
Hazard: Irritant
Formula:
C₁₂H₆F₃N₃O₂ Melting Pt: 230…233 °C |
MDL Number:
MFCD00203322 CAS Number: 75838-24-3 |
Specification Test Results
Melting point | 230-233°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...