trans-3,5-Difluorocinnamic acid ≥98%

Supplier: Matrix Scientific

Synonyms: trans-3-(3,5-Difluorophenyl)acrylic acid

001530-25G 001530-100G
101781-906EA 106.15 USD
101781-906 101781-908
trans-3,5-Difluorocinnamic acid ≥98%
trans-3,5-Difluorocinnamic acid

Hazard: Irritant

Formula: C₉H₆F₂O₂
MW: 184.14 g/mol
Melting Pt: 63-65°C
MDL Number: MFCD00010321
CAS Number: 147700-58-1

Order Now

Specification Test Results

Melting point 204-205°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR