4-O-Methyldopamine hydrochloride 99

Supplier: Matrix Scientific

001494-250MG 001494-1G
101781-810EA 250.77 USD
101781-810 101781-812
4-O-Methyldopamine hydrochloride 99
4-O-Methyldopamine hydrochloride

Hazard: Irritant

Formula: C₉H₁₄ClNO₂
MW: 203.666 g/mol
Melting Pt: 207…208 °C
MDL Number: MFCD00012896
CAS Number: 645-33-0

Order Now

Specification Test Results

Melting point 207-208°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR