2-Methoxy-4-(trifluoromethyl)pyridine-5-carboxamide ≥97%

Supplier: Matrix Scientific

001486-250MG 001486-1G
101781-798EA 35.38 USD
101781-798 101781-800
2-Methoxy-4-(trifluoromethyl)pyridine-5-carboxamide ≥97%
2-Methoxy-4-(trifluoromethyl)pyridine-5-carboxamide

Hazard: Irritant

Formula: C₈H₇F₃N₂O₂
Melting Pt: 182…184 °C
MDL Number: MFCD00173958
CAS Number: 175204-87-2

Order Now

Specification Test Results

Melting point 182-184°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR