4-Hydroxy-3-methylbenzoic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-m-toluic acid

001459-5G 001459-25G
101781-712EA 15.38 USD
101781-712 101781-714
4-Hydroxy-3-methylbenzoic acid ≥98%
4-Hydroxy-3-methylbenzoic acid

Purity: 98% CAS: 499-76-3 MDL Number: MFCD00270105 Molecular Formula: C8H8O3 Molecular Weight: 152,15 Hazard: Irritant

Formula: C₈H₈O₃
MW: 152.15 g/mol
Melting Pt: 174…175 °C
MDL Number: MFCD00270105
CAS Number: 499-76-3

Order Now

Specification Test Results

Melting point 174-175°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR