2-Hydroxy-3-methoxyphenylacetonitrile ≥98%
Supplier: Matrix Scientific
001457-1G
001457-5G
101781-704EA
96.92
USD
101781-704
101781-706
2-Hydroxy-3-methoxyphenylacetonitrile ≥98%
2-Hydroxy-3-methoxyphenylacetonitrile
Hazard: Irritant-Harmful
Formula:
C₉H₉NO₂ Melting Pt: 104-106°C |
CAS Number:
42973-56-8 |
Specification Test Results
Melting point | 104-106°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...