4-Hydroxy-2-(trifluoromethyl)pyrimidine

Supplier: Matrix Scientific

Synonyms: 2-(Trifluoromethyl)pyrimidin-4-ol

001424-1G 001424-5G
101781-628EA 64.62 USD
101781-628 101781-630
4-Hydroxy-2-(trifluoromethyl)pyrimidine
4-Hydroxy-2-(trifluoromethyl)pyrimidine

Hazard: Irritant

Formula: C₅H₃F₃N₂O
MW: 164.09 g/mol
Melting Pt: 168…171 °C
MDL Number: MFCD00151833
CAS Number: 1546-80-1

Order Now

Specification Test Results

Melting point 168-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR