[2-Nitro-4-(trifluoromethyl)phenyl]thioglycolic acid

Supplier: Matrix Scientific

001353-1G 001353-5G
101781-468EA 30.77 USD
101781-468 101781-470
[2-Nitro-4-(trifluoromethyl)phenyl]thioglycolic acid
[2-Nitro-4-(trifluoromethyl)phenyl]thioglycolic acid

Hazard: Irritant

Formula: C₉H₆F₃NO₄S
MW: 281.21 g/mol
Melting Pt: 156…158 °C
MDL Number: MFCD00177986
CAS Number: 728-56-3

Order Now

Specification Test Results

Melting point 156-158°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR