4'-Cyano-3'-(trifluoromethyl)acetanilide ≥98%

Supplier: Matrix Scientific

001346-5G 001346-25G
101781-446EA 26.15 USD
101781-446 101781-448
4'-Cyano-3'-(trifluoromethyl)acetanilide ≥98%
4'-Cyano-3'-(trifluoromethyl)acetanilide

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O
Melting Pt: 170-173°C
MDL Number: MFCD00052360
CAS Number: 97760-99-1

Order Now

Specification Test Results

Melting point 170-173°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR