4'-Cyano-2'-(trifluoromethyl)acetanilide ≥98%

Supplier: Matrix Scientific

001345-1G 001345-5G
101781-442EA 41.54 USD
101781-442 101781-444
4'-Cyano-2'-(trifluoromethyl)acetanilide ≥98%
4'-Cyano-2'-(trifluoromethyl)acetanilide

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O
Melting Pt: 169-171°C
MDL Number: MFCD00052844
CAS Number: 175277-96-0

Order Now

Specification Test Results

Melting point 169-171°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR