4'-Cyano-2'-(trifluoromethoxy)acetanilide ≥97%

Supplier: Matrix Scientific

001344-1G 001344-5G
101781-438EA 61.54 USD
101781-438 101781-440
4'-Cyano-2'-(trifluoromethoxy)acetanilide ≥97%
4'-Cyano-2'-(trifluoromethoxy)acetanilide

Hazard: Irritant

Formula: C₁₀H₇F₃N₂O₂
Melting Pt: 118-120°C
MDL Number: MFCD00204174
CAS Number: 175278-19-0

Order Now

Specification Test Results

Melting point 118-120°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR