4-Amino-3-(trifluoromethoxy)benzonitrile ≥97%
Supplier: Matrix Scientific
001332-1G
001332-5G
101781-412EA
53.85
USD
101781-412
101781-414
4-Amino-3-(trifluoromethoxy)benzonitrile ≥97%
4-Amino-3-(trifluoromethoxy)benzonitrile
Hazard: Irritant
Formula:
C₈H₅F₃N₂O MW: 202.1300 g/mol Melting Pt: 71…73 °C |
MDL Number:
MFCD00204177 CAS Number: 175278-23-6 |
Specification Test Results
Melting point | 71-73°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...