Ethyl-2,4,5-trifluorobenzoylacetate
Supplier: Matrix Scientific
Synonyms:
(2,4,5-Trifluorobenzoyl)acetic acid ethyl ester, 3-Oxo-3-(2,4,5-trifluorophenyl)propanoic acid ethyl ester, Ethyl (2,4,5-Trifluorobenzoyl)acetate, Ethyl 2,4,5-trifluorobenzoylacetate, Ethyl 3-Oxo-3-(2,4,5-trifluorophenyl)propanoate
001293-1G
001293-5G
101781-340EA
29.23
USD
101781-340
101781-342
Ethyl-2,4,5-trifluorobenzoylacetate
Ethyl-2,4,5-trifluorobenzoylacetate
Hazard: Irritant
Formula:
C₁₁H₉F₃O₃ MW: 246.19 g/mol Melting Pt: 66…68 °C |
MDL Number:
MFCD00792431 CAS Number: 98349-24-7 |
Specification Test Results
Melting point | 66-68°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...