(2-Bromo-4,5-dimethoxyphenyl)acetonitrile

Supplier: Matrix Scientific

Synonyms: 2-(2-Bromo-4,5-dimethoxyphenyl)acetonitrile

001291-1G 001291-5G
101781-332EA 32.31 USD
101781-332 101781-334
(2-Bromo-4,5-dimethoxyphenyl)acetonitrile
(2-Bromo-4,5-dimethoxyphenyl)acetonitrile

Hazard: Irritant/Irritant-Harmful

Formula: C₁₀H₁₀BrNO₂
MW: 256.1000 g/mol
Melting Pt: 87-89°C
MDL Number: MFCD00016385
CAS Number: 51655-39-1

Order Now

Specification Test Results

Melting point 87-89°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR