3-Amino-2,5,6-trifluorobenzoic acid ≥98%

Supplier: Matrix Scientific

001263-1G 001263-5G
101781-264EA 89.23 USD
101781-264 101781-266
3-Amino-2,5,6-trifluorobenzoic acid ≥98%
3-Amino-2,5,6-trifluorobenzoic acid

Hazard: Irritant

Formula: C₇H₄F₃NO₂
Melting Pt: 175-180°C
MDL Number: MFCD00075469
CAS Number: 133622-65-8

Order Now

Specification Test Results

Melting point 175-180°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR