(±)-3-Amino-4,4,4-trifluorobutyric acid ≥97%

Supplier: Matrix Scientific

001213-1G 001213-5G
101781-166EA 53.85 USD
101781-166 101781-168
(±)-3-Amino-4,4,4-trifluorobutyric acid ≥97%
(±)-3-Amino-4,4,4-trifluorobutyric acid

Hazard: Irritant

Formula: C₄H₆F₃NO₂
MW: 157.09
Melting Pt: 185…190 °C
MDL Number: MFCD00041413
CAS Number: 584-20-3

Order Now

Specification Test Results

Melting point 185-190°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR