4-Amino-2,3,5,6-tetrafluoropyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 2,3,5,6-Tetrafluoropyridin-4-amine

001177-5G 001177-25G
101781-090EA 73.85 USD
101781-090 101781-092
4-Amino-2,3,5,6-tetrafluoropyridine ≥98%
4-Amino-2,3,5,6-tetrafluoropyridine

Hazard: Irritant

Formula: C₅H₂F₄N₂
MW: 166.08 g/mol
Melting Pt: 85-87°C
MDL Number: MFCD00010855
CAS Number: 1682-20-8

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR