4'-Iodo-2'-(trifluoromethyl)acetanilide ≥97%
Supplier: Matrix Scientific
001065-1G
001065-5G
101780-898EA
73.85
USD
101780-898
101780-900
4'-Iodo-2'-(trifluoromethyl)acetanilide ≥97%
4'-Iodo-2'-(trifluoromethyl)acetanilide
Hazard: Irritant
Formula:
C₉H₇F₃INO Melting Pt: 134…136 °C |
MDL Number:
MFCD00052845 CAS Number: 97760-98-0 |
Specification Test Results
Melting point | 134-136°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...