2-Bromo-2',6'-dichloro-4'-(trifluoromethyl)acetophenone ≥95%

Supplier: Matrix Scientific

001063-100MG 001063-500MG
101780-892EA 93.85 USD
101780-892 101780-894
2-Bromo-2',6'-dichloro-4'-(trifluoromethyl)acetophenone ≥95%
2-Bromo-2',6'-dichloro-4'-(trifluoromethyl)acetophenone

Hazard: Irritant

Formula: C₉H₄BrCl₂F₃O
Melting Pt: 45…48 °C
MDL Number: MFCD00221016
CAS Number: 175205-89-7

Order Now

Specification Test Results

Melting point 45-48°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR