Ethyl-3-aminopropanoate hydrochloride ≥98%

Supplier: Matrix Scientific

001020-25G 001020-100G
101780-870EA 23.08 USD
101780-870 101780-872
Ethyl-3-aminopropanoate hydrochloride ≥98%
Ethyl-3-aminopropanoate hydrochloride

Hazard: Hygroscopic

Formula: C₅H₁₁NO₂·HCl
Melting Pt: 67…70 °C
MDL Number: MFCD00012909
CAS Number: 4244-84-2

Order Now

Specification Test Results

Melting point 70-72°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR