2-Formyl-6-picoline ≥98%

Supplier: Matrix Scientific

Synonyms: 6-Methylpyridine-2-carboxaldehyde, 6-Methyl-2-pyridinecarboxaldehyde

012634-5G 012634-25G
101803-040EA 44.62 USD
101803-040 101803-042
2-Formyl-6-picoline ≥98%
2-Formyl-6-picoline

TCA listed: T
Hazard: Irritant/Light Sensitive/Store in Freezer

Formula: C₇H₇NO
MW: 121.14 g/mol
Boiling Pt: 77…78 °C (12 Torr)
Melting Pt: 31…33 °C
MDL Number: MFCD00006291
CAS Number: 1122-72-1

Order Now

Specification Test Results

Melting point 31-33°C
ND20 1.527


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR