Isopropyl anthranilate ≥99%
Supplier: Matrix Scientific
001016-5G
001016-25G
101780-864EA
73.85
USD
101780-864
101780-866
Isopropyl anthranilate ≥99%
Isopropyl anthranilate
Hazard code(s): Irritant
Formula:
C₁₀H₁₃NO₂ Melting Pt: 64…65 °C |
MDL Number:
MFCD00060604 CAS Number: 18189-02-1 EINECS: 242-074-9 |
Specification Test Results
Melting point | 64-65°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...