6-Chloro-2-methoxy-3-pyridineboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: (6-Chloro-2-methoxypyridin-3-yl)boronic acid

070539-1G 070539-5G
101945-214EA 233.85 USD
101945-214 101945-216
6-Chloro-2-methoxy-3-pyridineboronic acid ≥97%
6-Chloro-2-methoxy-3-pyridineboronic acid

Hazard code(s): Irritant

Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
MDL Number: MFCD11053817
CAS Number: 1072946-50-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR