2,4,6-Tribromopyrimidine ≥97%

Supplier: Matrix Scientific
Warning

070426-1G 070426-5G
101944-698EA 524.62 USD
101944-698 101944-700
2,4,6-Tribromopyrimidine ≥97%
2,4,6-Tribromopyrimidine

Hazard code(s): Irritant

Formula: C₄HBr₃N₂
MW: 316.78
MDL Number: MFCD00233942
CAS Number: 36847-11-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR