5-Chloro-2-fluorobenzaldehyde ≥97%

Supplier: Matrix Scientific

070410-5G 070410-25G
101944-626EA 15.38 USD
101944-626 101944-628
5-Chloro-2-fluorobenzaldehyde ≥97%
5-Chloro-2-fluorobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄ClFO
MDL Number: MFCD03094518
CAS Number: 96515-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR