Tributylvinyltin ≥97%

Supplier: Matrix Scientific
Danger

Synonyms: Tri-n-butylvinyltin, Tributylvinylstannane

070383-5G 070383-25G
101944-506EA 60 USD
101944-506 101944-508
Tributylvinyltin ≥97%
Tributylvinyltin

Hazard code(s): Toxic, Marine Pollutant

Formula: C₁₄H₃₀Sn
MW: 317.1 g/mol
Boiling Pt: 104…106 °C (3.5mm)
Density: 1.085 g/cm³ (25 °C)
MDL Number: MFCD00009421
CAS Number: 7486-35-3
EINECS: 231-291-4
UN: 2788
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR