Tributylallyltin ≥97% for synthesis

Supplier: Matrix Scientific
Danger

Synonyms: Allyltributylstannane, Allyltributyltin, Tributylallylstannane, Tributyl-2-propenylstannane, Allyltri-n-butyltin, Tri-n-butyl-2-propenylstannane, Allyltri-n-butylstannane

070382-25G 070382-100G
101944-502EA 66.15 USD
101944-502 102833-478
Tributylallyltin ≥97% for synthesis
Tributylallyltin

Hazard code(s): Toxic, Air Sensitive

Formula: C₁₅H₃₂Sn
Boiling Pt: 100…102 °C (0.5mm)
Density: 1.073 g/cm³ (25 °C)
MDL Number: MFCD00010346
CAS Number: 24850-33-7
EINECS: 246-494-3
UN: 2788
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR