3-Bromobiphenyl (PBB No. 2) ≥98%

Supplier: Matrix Scientific

Synonyms: PBB No. 2

070380-5G 070380-25G 070380-100G
101944-486EA 46.15 USD
101944-486 101944-488 101944-490
3-Bromobiphenyl (PBB No. 2) ≥98%
3-Bromobiphenyl (PBB No. 2)

Hazard code(s): Irritant

Formula: C₁₂H₉Br
MW: 233.11 g/mol
Boiling Pt: 299 °C (1013 hPa)
Melting Pt: 9 °C
Density: 1.41 g/cm³ (20 °C)
MDL Number: MFCD00000082
CAS Number: 2113-57-7
EINECS: 218-304-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR